Anti-Vaccinia virus free virus Mouse Monoclonal Antibody [clone: 8112]

Supplier: GeneTex
GTX36669
89331-728EA 652.21 USD
89331-728
Anti-Vaccinia virus free virus Mouse Monoclonal Antibody [clone: 8112]
Antibodies
Type: Primary
Antigen: Vaccinia virus free virus
Clonality: Monoclonal
Clone: 8112
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR