Anti-Vaccinia virus free virus Mouse Monoclonal Antibody [clone: 8116]

Supplier: GeneTex
GTX36667
89331-724EA 652.21 USD
89331-724
Anti-Vaccinia virus free virus Mouse Monoclonal Antibody [clone: 8116]
Antibodies
Type: Primary
Antigen: Vaccinia virus free virus
Clonality: Monoclonal
Clone: 8116
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR