Anti-Vaccinia virus free virus Mouse Monoclonal Antibody [clone: 8118]

Supplier: GeneTex
GTX36666
89331-722EA 652.21 USD
89331-722
Anti-Vaccinia virus free virus Mouse Monoclonal Antibody [clone: 8118]
Antibodies
Type: Primary
Antigen: Vaccinia virus free virus
Clonality: Monoclonal
Clone: 8118
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR