Anti-Vaccinia virus Mouse Monoclonal Antibody [clone: 8119]

Supplier: GeneTex
GTX36665
89331-720EA 652.21 USD
89331-720
Anti-Vaccinia virus Mouse Monoclonal Antibody [clone: 8119]
Antibodies
Type: Primary
Antigen: Vaccinia virus
Clonality: Monoclonal
Clone: 8119
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR