Anti-HSV1 Mouse Monoclonal Antibody [clone: 112]

Supplier: GeneTex
GTX36663
89331-716EA 652.21 USD
89331-716
Anti-HSV1 Mouse Monoclonal Antibody [clone: 112]
Antibodies
Type: Primary
Antigen: HSV1
Clonality: Monoclonal
Clone: 112
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR