Anti-HSV2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 111]

Supplier: GeneTex
GTX36662
89331-714EA 651.82 USD
89331-714
Anti-HSV2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 111]
Antibodies
Type: Primary
Antigen: HSV2
Clonality: Monoclonal
Clone: 111
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR