Anti-HSV2 Mouse Monoclonal Antibody [clone: 116]

Supplier: GeneTex
GTX36660
89331-710EA 652.21 USD
89331-710
Anti-HSV2 Mouse Monoclonal Antibody [clone: 116]
Antibodies
Type: Primary
Antigen: HSV2
Clonality: Monoclonal
Clone: 116
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR