Anti-HSV1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 181]

Supplier: GeneTex
GTX36653
89331-700EA 395.14 USD
89331-700
Anti-HSV1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 181]
Type: Primary
Antigen: HSV1
Clonality: Monoclonal
Clone: 181
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR