Anti-HSV1/2 Mouse Monoclonal Antibody [clone: 195]

Supplier: GeneTex
GTX36650
89331-694EA 539.43 USD
89331-694
Anti-HSV1/2 Mouse Monoclonal Antibody [clone: 195]
Antibodies
Type: Primary
Antigen: HSV1/2
Clonality: Monoclonal
Clone: 195
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR