Anti-Epstein-Barr virus gp 250/350 Mouse Monoclonal Antibody [clone: 221]

Supplier: GeneTex
GTX36648
89331-692EA 652.21 USD
89331-692
Anti-Epstein-Barr virus gp 250/350 Mouse Monoclonal Antibody [clone: 221]
Antibodies
Type: Primary
Antigen: Epstein-Barr virus gp 250/350
Clonality: Monoclonal
Clone: 221
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR