Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Epstein-Barr virus gp 250/350 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 221]

Supplier: Genetex

GTX36647
89331-690EA 359.03 USD InStock
89331-690
Anti-Epstein-Barr virus gp 250/350 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 221]
Antibodies
Type: Primary
Antigen: Epstein-Barr virus gp 250/350
Clonality: Monoclonal
Clone: 221
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty