Anti-Epstein-Barr virus VCA Mouse Monoclonal Antibody [clone: 231]

Supplier: GeneTex
GTX36646
89331-688EA 539.43 USD
89331-688
Anti-Epstein-Barr virus VCA Mouse Monoclonal Antibody [clone: 231]
Antibodies
Type: Primary
Antigen: Epstein-Barr virus VCA
Clonality: Monoclonal
Clone: 231
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR