Anti-Epstein-Barr virus Bam H1 Z Mouse Monoclonal Antibody [clone: 256]

Supplier: GeneTex
GTX36643
89331-684EA 652.21 USD
89331-684
Anti-Epstein-Barr virus Bam H1 Z Mouse Monoclonal Antibody [clone: 256]
Antibodies
Type: Primary
Antigen: Epstein-Barr virus Bam H1 Z
Clonality: Monoclonal
Clone: 256
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR