Anti-Streptococcus pneumoniae Mouse Monoclonal Antibody [clone: 471]

Supplier: GeneTex
GTX36636
89331-670EA 652.21 USD
89331-670
Anti-Streptococcus pneumoniae Mouse Monoclonal Antibody [clone: 471]
Antibodies
Type: Primary
Antigen: Streptococcus pneumoniae
Clonality: Monoclonal
Clone: 471
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR