Anti-Rotavirus capsid Mouse Monoclonal Antibody [clone: 581]

Supplier: GeneTex
GTX36628
89331-656EA 468.76 USD
89331-656
Anti-Rotavirus capsid Mouse Monoclonal Antibody [clone: 581]
Antibodies
Type: Primary
Antigen: Rotavirus capsid
Clonality: Monoclonal
Clone: 581
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR