Anti-Cytomegalovirus gB Mouse Monoclonal Antibody [clone: 826]

Supplier: GeneTex
GTX36617
89331-640EA 652.21 USD
89331-640
Anti-Cytomegalovirus gB Mouse Monoclonal Antibody [clone: 826]
Antibodies
Type: Primary
Antigen: Cytomegalovirus gB
Clonality: Monoclonal
Clone: 826
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR