Anti-Shigella dysenteriae Mouse Monoclonal Antibody [clone: 911]

GTX36606
89331-618EA 377.13 USD
89331-618
Anti-Shigella dysenteriae Mouse Monoclonal Antibody [clone: 911]
Type: Primary
Antigen: Shigella dysenteriae
Clonality: Monoclonal
Clone: 911
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR