Anti-Respiratory Syncytial virus Goat Polyclonal Antibody

Supplier: GeneTex
GTX36603
89331-616EA 666.59 USD
89331-616
Anti-Respiratory Syncytial virus Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Respiratory Syncytial virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR