Anti-HSV2 Sheep Polyclonal Antibody

GTX36598
89331-606EA 376.67 USD
89331-606
Anti-HSV2 Sheep Polyclonal Antibody
Type: Primary
Antigen: HSV2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR