Anti-Hepatitis B santigen ad/ay Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36575
89331-560EA 660.25 USD
89331-560
Anti-Hepatitis B santigen ad/ay Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Hepatitis B santigen ad/ay
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR