Anti-Pichia pastoris Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36555
89331-520EA 660.25 USD
89331-520
Anti-Pichia pastoris Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Pichia pastoris
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR