Anti-Tetanus toxoid Goat Polyclonal Antibody

Supplier: GeneTex
GTX36553
89331-516EA 683.75 USD
89331-516
Anti-Tetanus toxoid Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Tetanus toxoid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR