Anti-Hepatitis A virus VP3 Goat Polyclonal Antibody

GTX36543
89331-496EA 377.16 USD
89331-496
Anti-Hepatitis A virus VP3 Goat Polyclonal Antibody
Type: Primary
Antigen: Hepatitis A virus VP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR