Anti-NS4 Goat Polyclonal Antibody

GTX36539
89331-488EA 376.45 USD
89331-488
Anti-NS4 Goat Polyclonal Antibody
Type: Primary
Antigen: NS4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR