Anti-Streptococcus, group A Rabbit Polyclonal Antibody

GTX36530
89331-474EA 376.66 USD
89331-474
Anti-Streptococcus, group A Rabbit Polyclonal Antibody
Type: Primary
Antigen: Streptococcus, group A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR