Anti-HSV1/2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36523
89331-462EA 652.02 USD
89331-462
Anti-HSV1/2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSV1/2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR