Anti-HSV2 Sheep Polyclonal Antibody

Supplier: GeneTex
GTX36522
89331-460EA 656.63 USD
89331-460
Anti-HSV2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSV2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR