Anti-Rubella virus virions Goat Polyclonal Antibody

Supplier: GeneTex
GTX36513
89331-442EA 656.63 USD
89331-442
Anti-Rubella virus virions Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Rubella virus virions
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR