Anti-Helicobacter pylori Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36493
89331-404EA 652.21 USD
89331-404
Anti-Helicobacter pylori Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Helicobacter pylori
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR