Anti-E. coli Goat Polyclonal Antibody

Supplier: GeneTex
GTX36484
89331-388EA 656.44 USD
89331-388
Anti-E. coli Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: E. coli
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR