Anti-CTB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36480
89331-380EA 652.21 USD
89331-380
Anti-CTB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CTB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR