Anti-E. coli Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36461
89331-364EA 358 USD
89331-364
Anti-E. coli Rabbit Polyclonal Antibody
Type: Primary
Antigen: E. coli
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR