Anti-Parainfluenza types 2 & 3 Goat Polyclonal Antibody

Supplier: GeneTex
GTX36444
89331-334EA 660.07 USD
89331-334
Anti-Parainfluenza types 2 & 3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Parainfluenza types 2 & 3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR