Anti-Hepatitis B santigen ad/ay Goat Polyclonal Antibody

GTX36436
89331-318EA 376.67 USD
89331-318
Anti-Hepatitis B santigen ad/ay Goat Polyclonal Antibody
Type: Primary
Antigen: Hepatitis B santigen ad/ay
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR