Anti-Leptospira biflexa Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36427
89331-302EA 660.25 USD
89331-302
Anti-Leptospira biflexa Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Leptospira biflexa
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR