Anti-Candida albicans Rabbit Polyclonal Antibody

GTX36425
89331-298EA 376.67 USD
89331-298
Anti-Candida albicans Rabbit Polyclonal Antibody
Type: Primary
Antigen: Candida albicans
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR