Anti-Campylobacter jejuni Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36413
89331-276EA 660.25 USD
89331-276
Anti-Campylobacter jejuni Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Campylobacter jejuni
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR