Anti-Neisseria meningitids Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36405
89331-262EA 660.25 USD
89331-262
Anti-Neisseria meningitids Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Neisseria meningitids
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR