Anti-Epstein-Barr virus EBNA-1 Goat Polyclonal Antibody

GTX36400
89331-258EA 376.99 USD
89331-258
Anti-Epstein-Barr virus EBNA-1 Goat Polyclonal Antibody
Type: Primary
Antigen: Epstein-Barr virus EBNA-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR