Anti-Clostridia Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36392
89331-246EA 660.25 USD
89331-246
Anti-Clostridia Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Clostridia
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR