Anti-Enterococcus sp. Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36383
89331-240EA 660.25 USD
89331-240
Anti-Enterococcus sp. Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Enterococcus sp.
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR