Anti-Parainfluenza types 2 & 3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

GTX36368
89331-214EA 376.99 USD
89331-214
Anti-Parainfluenza types 2 & 3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: Parainfluenza types 2 & 3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR