Anti-Vaccinia virus Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

GTX36350
89331-180EA 376.99 USD
89331-180
Anti-Vaccinia virus Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: Vaccinia virus
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR