Anti-Hepatitis B santigen ad/ay Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX36347
89331-174EA 659.35 USD
89331-174
Anti-Hepatitis B santigen ad/ay Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: Hepatitis B santigen ad/ay
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR