Anti-Diphtheria toxin Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

GTX36336
89331-152EA 376.67 USD
89331-152
Anti-Diphtheria toxin Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: Diphtheria toxin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR