Anti-Streptococcus, group A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX36302
89331-114EA 631.75 USD
89331-114
Anti-Streptococcus, group A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: Streptococcus, group A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR