Anti-FLT3 Rabbit Monoclonal Antibody [clone: T.602.6]

Supplier: GeneTex
GTX36299
89304-024EA 610.26 USD
89304-024
Anti-FLT3 Rabbit Monoclonal Antibody [clone: T.602.6]
Antibodies
Rabbit monoclonal antibody [T.602.6] to FLT3 (phospho Tyr969)

Recommended Starting Dilutions:WB: 1:1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: FLT3
Clonality: Monoclonal
Clone: T.602.6
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR