Anti-Anopheles stephensi protein 1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

GTX36292
89276-432EA 473.12 USD
89276-432
Anti-Anopheles stephensi protein 1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Primary
Antigen: Anopheles stephensi protein 1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR