Anti-FTSZ Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX36253
89284-690EA 615.79 USD
89284-690
Anti-FTSZ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FtsZ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacillus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR