Anti-IgA Mouse Monoclonal Antibody (Biotin)

Supplier: GeneTex
GTX35079
89306-988EA 129.57 USD
89306-988
Anti-IgA Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR